Learn More
ACROBiosystems Human S100A14 Protein, His Tag
Marke: ACROBiosystems S14-H5121-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Protein S100-A14 (S100A14) is also known as S100 calcium-binding protein A14 (S114),which belongs to the S-100 family, containing one EF-hand domain.S100A14 is a homodimer can interact with AGER. S100A14 can modulate P53/TP53 protein levels, and play a role in the regulation of cell survival and apoptosis at different concentrations via receptor for advanced glycation end products (RAGE).S100A14 also plays a role in the regulation of cell migration by modulating the levels of MMP2, a matrix protease that is under transcriptional control of P53/TP53.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.5 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH05019 | |
100 μg | |
Met 1 - His 104 | |
S100A14,S114,S100A15 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
12.5 kDa | |
Human S100A14, His Tag (S14-H5121) is expressed from E.coli cells. It contains AA Met 1 - His 104 (Accession # Q9HCY8-1). | |
-20°C | |
S100A14 | |
Human | |
50 mM Tris, 150 mM NaCl, pH8.0 | |
E.coli | |
Other Recombinant Proteins |