Learn More
ACROBiosystems Human VEGF-C / Flt4-L Protein, His Tag
Brand: ACROBiosystems VEC-H4225-50UG
Additional Details : Netto-Gewicht : 0.00001kg
Description
Vascular endothelial growth factor C is also known as VEGFC, Flt4-L and VRP, it contains the C-terminal propeptide which has an unusual structure with tandemly repeated cysteine-rich motifs. Upon biosynthesis, VEGFC is secreted as a non-covalent momodimer in an anti-parellel fashion. VEGFC is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis, lymphangiogenesis and endothelial cell growth and survival, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms that bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. The structure and function of this protein is similar to those of vascular endothelial growth factor D (VEGF-D). VEGFC may function in angiogenesis of the venous and lymphatic vascular systems during embryogenesis, and also in the maintenance of differentiated lymphatic endothelium in adults. Overexpression of VEGF-C causes lymphatics to enlarge possibly facilitates metastasis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 19-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH35212.1 | |
50 μg | |
Thr 103 - Arg 227 | |
VEGFC,Flt4-L,VRP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
14.9 kDa | |
Human VEGF-C, His Tag (VEC-H4225) is expressed from human 293 cells (HEK293). It contains AA Thr 103 - Arg 227 (Accession # AAH35212.1). | |
-20°C | |
VEGF-C | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |