Learn More
ACROBiosystems Human CD9 Protein, His Tag
Marke: ACROBiosystems CD9-H5220-20UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
CD9 antigen is also known as tetraspanin-29 (TSPAN29), 5H9 antigen, Leukocyte antigen MIC3 (MIC3), Motility-related protein, is a multi-pass membrane protein which belongs to the tetraspanin (TM4SF) family or the transmembrane 4 superfamily. CD9 is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. TSPAN29 is found on the surface of exosomes. MIC3 Involved in platelet activation and aggregation, regulates paranodal junction formation and also Involved in cell adhesion, cell motility and tumor metastasis. CD9 antigen also seems to be a key part in the egg-sperm fusion during mammalian fertilization.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.6 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
NP_001760.1 | |
20 μg | |
Ser 112 - Ile 195 | |
CD9,MIC3,TSPAN29,GIG2,MRP1,BTCC1,DRAP27,5H9 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
11.6 kDa | |
Human CD9, His Tag (CD9-H5220) is expressed from human 293 cells (HEK293). It contains AA Ser 112 - Ile 195 (Accession # P21926-1). | |
-20°C | |
CD9 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |