Learn More
ACROBiosystems Human CEACAM-8 / CD66b Protein, His Tag
Marke:  ACROBiosystems CE8-H5224-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15856437
  • 395.89 € / 100 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Carcinoembryonic antigen-related cell adhesion molecule 8 (CEACAM8) is also known as CD66b (Cluster of Differentiation 66b), CD66b, CD67, CGM6, NCA-95, and is one of seven human CEACAM family members within the immunoglobulin superfamily. CEACAM family members are a set of widely expressed proteins involved in several biological functions, including cell adhesion, migration, signal transduction, and the regulation of gene expression. Abnormal overexpression and downregulation of some CEACAMs have been described in tumor cells. In humans, CEACAMs include type I transmembrane proteins (CEACAM1, CEACAM3, and CEACAM4) and GPI-linked molecules (CEACAM5 through CEACAM8). There is no human CEACAM2. CEACAM8 is a single chain, two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It is an activation marker for human granulocytes.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.2 kDa. The protein migrates as 53-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_001807.2
100 μg
Gln 35 - Ser 319
CEACAM8,CD66b,CD67,CGM6,NCA-95
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
32.2 kDa
Human CEACAM-8, His Tag (CE8-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Ser 319 (Accession # NP_001807.2).
-20°C
CEACAM-8
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens