Learn More
ACROBiosystems Human DLL4 Protein, His Tag
Marke:  ACROBiosystems DL4-H5227-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15856577
  • 299.92 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Delta-like protein 4 (DLL4) is also known as Drosophila Delta homolog 4 (Delta4), which contains one DSL domain and eight EGF-like domains. DLL4 is expressed in vascular endothelium. DLL4 is involved in the Notch signaling pathway as Notch ligand, which can activates NOTCH1 and NOTCH4. DLL4 is involved in angiogenesis and negatively regulates endothelial cell proliferation and migration and angiogenic sprouting. DLL4 can bind to Notch-1 and Notch-4.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.5 kDa. The protein migrates as 58-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAI06951
100 μg
Ser 27 - Pro 524
DLL4,Delta4
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
55.5 kDa
Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # Q9NR61-1).
-20°C
DLL4
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens