Learn More
ACROBiosystems Human IGFBP-7 Protein, His Tag
Marke: ACROBiosystems IG7-H5240-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Insulin-like growth factor-binding protein 7 (IGFBP7) is also known as IGFBP-rP1, MAC25 protein, PGI2-stimulating factor, prostacyclin-stimulating factor and tumor-derived adhesion factor, which contains one Ig-like C2-type (immunoglobulin-like) domain, one IGFBP N-terminal domain and one Kazal-like domain. The major function of IGFBP7 is the regulation of availability of insulin-like growth factors (IGFs) in tissue as well as in modulating IGF binding to its receptors. IGFBP7 binds to IGF with high affinity except for IGF-I and IGF-II.IGFBP7 also stimulates cell adhesion. Furthermore, IGFBP7 is implicated in some cancers.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27.0 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001544 | |
50 μg | |
Asp 30 - Leu 282 | |
IBP7,IGFBP7 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
27.0 kDa | |
Human IGFBP-7, His Tag (IG7-H5240) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Leu 282 (Accession # NP_001544). | |
-20°C | |
IGFBP-7 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |