Learn More
ACROBiosystems Human Integrin alpha 8 beta 1 (ITGA8&ITGB1) Heterodimer Protein, His Tag&Tag Free
Marke:  ACROBiosystems IT1-H52W9-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15857147
  • 491.86 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Human Integrin alpha 8 beta 1 Heterodimer Protein consists of ITGA8 and ITGB1. The integrin alpha 8 subunit, isolated by low stringency hybridization, is a novel integrin subunit that associates with beta 1. The recently identified alpha 8 integrin subunit associates with beta 1 and is predominantly expressed in smooth muscle and other contractile cells in adult tissues, and in mesenchymal and neural cells during development. In addition, Integrin alpha 8 beta 1 is a receptor for fibronectin and can promote attachment, cell spreading, and neurite outgrowth on fibronectin.
Human ITGA8&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA8 and ITGB1, has a calculated MW of 114.6 kDa (ITGA8) and 83.7 kDa (ITGB1). Subunit ITGA8 is fused with an acidic tail at the C-terminus and followed by a polyhistid
Spezifikation
Spezifikation
NP 003629.2 (ITGA8) and NP_002202.2 (ITGB1)
100 μg
Phe 39 - Leu 1012 (ITGA8) and Gln 21 - Asp 728 (ITGB1)
Integrin alpha 8 beta 1,ITGA8andITGB1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
114.6 kDa (ITGA8) and 83.7 kDa (ITGB1)
Human ITGA8&ITGB1; Heterodimer Protein, His Tag&Tag; Free (IT1-H52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 39 - Leu 1012 (ITGA8) and Gln 21 - Asp 728 (ITGB1) (Accession # P53708-1 (ITGA8) and P05556-1 (ITGB1)).
-20°C
Integrin alpha 8 beta 1
Human
50 mM Tris, 150 mM NaCl, pH7.5
HEK293
Bio-Markers and CD Antigens