Learn More
ACROBiosystems Biotinylated Human Mesothelin / MSLN (296-580) Protein, His Tag, ultra sensitivity (primary amine labeling)
Marke:  ACROBiosystems MSN-H8223-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15857327
  • 347.90 € / 25 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.0 kDa. The protein migrates as 38-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH09272
25 μg
Glu 296 - Gly 580
MSLN,Mesothelin,MPF
UltraLys
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
33.0 kDa
Biotinylated Human Mesothelin (296-580), His Tag, primary amine labeling (MSN-H8223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272). It is the biotinylated form of Human Mesothelin (296-580), His Tag (MSN-H5223).
-20°C
Mesothelin
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins