Learn More
ACROBiosystems Human SOD1 / Cu-Zn SOD Protein, His Tag
Marke:  ACROBiosystems SO1-H5148-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15857717
  • 239.93 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Superoxide dismutase [Cu-Zn] (SOD1) is also known as superoxide dismutase 1 (hSod1), an enzyme that in humans is encoded by the SOD1 gene, located on chromosome 21. SOD1 can bind copper and zinc ions and is one of three superoxide dismutases responsible for destroying free superoxide radicals in the body. The encoded isozyme (SOD1) is a soluble cytoplasmic and mitochondrial intermembrane space protein, acting as a homodimer to convert naturally occurring, but harmful, superoxide radicals to molecular oxygen and hydrogen peroxide. Furthermore, the mutations of SOD1 gene can result in a neurodegenerative disorder affecting upper motor neurons in the brain and lower motor neurons in the brain stem and spinal cord.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 20 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation
Spezifikation
AAH01034
100 μg
Ala 2 - Gln 154
SOD1,Superoxide dismutase 1,hSod1
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>97% as determined by SDS-PAGE
16.8 kDa
Human SOD1, His Tag (SO1-H5148) is expressed from E.coli cells. It contains AA Ala 2 - Gln 154 (Accession # P60052-1).
-20°C
SOD1
Human
50 mM Tris, 150 mM NaCl, pH7.5
E.coli
Other Recombinant Proteins