Learn More
ACROBiosystems Human VEGF165 Protein, His Tag
Marke: ACROBiosystems VE5-H5248-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
VEGF165 is the most abundant splice variant of VEGF-A. VEGF165 is produced by a number of cells including endothelial cells, macrophages and T cells. VEGF165 is involved in angiogenesis, vascular endothelial cell survival, growth, migration and vascular permeability. VEGF gene expression is induced by hypoxia, inflammatory cytokines and oncogenes. VEGF165 binds to heparan sulfate and is retained on the cell surface and in the extracellular matrix. VEGF165 binds to the receptor tyrosine kinases, VEGFR1 and VEGFR2. VEGF165 is the only splice variant that binds to co-receptors NRP-1 and NRP-2 that function to enhance VEGFR2 signaling. Binding of VEGF165 to VEGFR1 and VEGFR2 leads to activation of the PI3K/AKT, p38 MAPK, FAK and paxillin. VEGF plays a key role in tumor angiogenesis in many cancers.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 25-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001165097.1 | |
1 mg | |
Ala 27 - Arg 191 | |
RP1-261G23.1,MGC70609,MVCD1,VEGFA,VPF | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
21.1 kDa | |
Human VEGF165, His Tag (VE5-H5248) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4). | |
-20°C | |
VEGF165 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |