Learn More
ACROBiosystems Human ANGPTL7 / CDT6 Protein, His Tag
Marke: ACROBiosystems AN7-H52H5-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Angiopoietin-like 7 (ANGPTL7) is also known as Corneal-Derived Transcript 6 (CDT6), and is a secreted glycoprotein that is structurally related to the angiopoietins. It is expressed in the corneal stroma, trabecular meshwork, and sclera and is elevated in glaucoma aqueous humor. Recently, ANGPTL proteins, including ANGPTL-2, -3, -4, -5 and -7, are identified as growth factors in combination with SCF, Thrombopoietin, IGF-II and FGF acidic that enhance the expansion and engraftment of human and mouse hematopoietic stem cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.0 kDa. The protein migrates as 38-40 kDa and 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_066969.1 | |
100 μg | |
Gln 27 - Pro 346 | |
ANGPTL7,CDT6 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |
39.0 kDa | |
Human ANGPTL7, His Tag (AN7-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Pro 346 (Accession # O43827-1). | |
-20°C | |
ANGPTL7 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |