missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus / Rhesus macaque B7-2 / CD86 Protein, Fc Tag

Marke:  ACROBiosystems CD6-C5254-100UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15866217

  • 311.92 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Cluster of Differentiation 86 (CD86) is also known as B-lymphocyte activation antigen B7-2, is a type I membrane protein that is a member of the immunoglobulin superfamily, and is constitutively expressed on interdigitating dendritic cells, Langerhans cells, peripheral blood dendritic cells, memory B cells, and germinal center B cells. Additionally, B72 is expressed at low levels on monocytes and can be upregulated through interferon γ. CD86 is the ligand for two different proteins on the T cell surface: CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation). CD86 works in tandem with CD80 to prime T cells. Recent study has revealed that B7-2 promotes the generation of a mature APC repertoire and promotes APC function and survival. Furthermore, the B7 proteins are also involved in innate immune responses by activating NF-κB-signaling pathway in macrophages. CD86 thus is regarded as a promising candidate for immune therapy. CD86+ macrophages in Hodgkin lymphoma patients are an independent marker for potential nonresponse to firstline-therapy.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

52 kDa
Cynomolgus / Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession # G7NXR4). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7-2 are homologus.
-20°C
B7-2
Cynomolgus Monkey, Rhesus Macaque
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Immune Checkpoint Proteins
100 μg
Ala 19 - His 240
CD86,B7-2,B70,CD28LG2,LAB72,MGC34413
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote