Learn More
ACROBiosystems Biotinylated Human B7-2 / CD86 Protein, Avitag™,His Tag
Biotinylated Protein
Marke: ACROBiosystems CD6-H82E2-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Cluster of Differentiation 86 (CD86) is also known as B-lymphocyte activation antigen B7-2, is a type I membrane protein that is a member of the immunoglobulin superfamily, and is constitutively expressed on interdigitating dendritic cells, Langerhans cells, peripheral blood dendritic cells, memory B cells, and germinal center B cells. Additionally, B72 is expressed at low levels on monocytes and can be upregulated through interferon γ. CD86 is the ligand for two different proteins on the T cell surface: CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation). CD86 works in tandem with CD80 to prime T cells. Recent study has revealed that B7-2 promotes the generation of a mature APC repertoire and promotes APC function and survival. Furthermore, the B7 proteins are also involved in innate immune responses by activating NF-κB-signaling pathway in macrophages. CD86 thus is regarded as a promising candidate for immune therapy. CD86+ macrophages in Hodgkin lymphoma patients are an independent marker for potential nonresponse to firstline-therapy.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 28.0 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH40261.1 | |
25 μg | |
Leu 26 - Pro 247 | |
CD86,B7-2,B70,CD28LG2,LAB72,MGC34413 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
28.0 kDa | |
Biotinylated Human B7-2, Avitag,His Tag (CD6-H82E2) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession # AAH40261.1). | |
-20°C | |
B7-2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |