Learn More
ACROBiosystems Human CD3 delta Protein, His Tag
Marke:  ACROBiosystems CDD-H5224-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15866377
  • 387.09 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
CD3D is also known as CD3-DELTA, T3D, and is a single-pass type I membrane protein. CD3D is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The encoded membrane protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in this gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms have been found for this gene. Other variants may also exist, but the full-length natures of their transcripts has yet to be defined. Defects in CD3D cause severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (T-/B+/NK+ SCID) which is a genetically and clinically heterogeneous group of rare congenital disorders characterized by impairment of both humoral and cell-mediated immunity, leukopenia, and low or absent antibody levels.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.4 kDa. The protein migrates as 18-28 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation
Spezifikation
NP_000723.1
100 μg
Phe 22 - Ala 105
CD3D,CD3-DELTA,T3D
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
11.4 kDa
Human CD3 delta, His Tag (CDD-H5224) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession # P04234-1).
-20°C
CD3 delta
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens