Learn More
ACROBiosystems Biotinylated Human CD3 epsilon Protein, His Tag, ultra sensitivity (primary amine labeling)
Marke:  ACROBiosystems CDE-H8223-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15866397
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
CD3e molecule, epsilon is also known as CD3E, is a T-cell surface single-pass type I membrane glycoprotein. CD3E contains 1 Ig-like (immunoglobulin-like) domain and 1 ITAM domain. CD3E, together with CD3-gamma, CD3-delta and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. The epsilon polypeptide plays an essential role in T-cell development. CD3E plays an essential role in T-cell development, and defects in CD3E gene cause severe immunodeficiency. CD3E gene has also been linked to a susceptibility to type I diabetes in women. CD3E has been shown to interact with TOP2B, CD3EAP and NCK2.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.6 kDa. The protein migrates as 15-16 kDa under reducing (R) condition (SDS-PAGE).
Spezifikation
Spezifikation
NP_000724.1
25 µg
Asp 23 - Asp 126
FLJ18683,T3E,TCRE,CD3E,CD3-epsilon
UltraLys
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
>90% as determined by reduced SDS-PAGE
12.6 kDa
MABSol™ Biotinylated Human CD3E, His Tag, primary amine labeling (CDE-H8223) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (Accession # NP_000724.1). It is the biotinylated form of Human CD3 epsilon Protein (Cat # CDE-H5223).
-20°C
CD3 epsilon
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins