Learn More
ACROBiosystems Biotinylated Human CX3CL1 Protein, His Tag, ultra sensitivity (primary amine labeling)
Marke:  ACROBiosystems CX1-H8221-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15866547
  • 1847.47 € / 200 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Fractalkine is also known as C-X3-C motif chemokine 1 (CX3CL1) and neurotactin. Fractalkine is a large cytokine protein of 373 amino acids, it contains multiple domains and is the only known member of the CX3C chemokine family. Fractalkine is found commonly throughout the brain, particularly in neural cells, and its receptor is known to be present on microglial cells. It has also been found to be essential for microglial cell migration. CX3CL1 is also up-regulated in the hippocampus during a brief temporal window following spatial learning, the purpose of which may be to regulate glutamate-mediated neurotransmission tone. This indicates a possible role for the chemokine in the protective plasticity process of synaptic scaling.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa. The protein migrates as 50-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH01163
200 μg
Gln 25 - Gln 341
CX3CL1,Fractalkine
UltraLys
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
35.5 kDa
Biotinylated Human C x 3 CL1, His Tag, primary amine labeling (C x 1-H8221) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 341 (Accession # AAH01163). It is the biotinylated form of Human C x 3 CL1, His Tag (Cat. No.C x 1-H5221).
-20°C
CX3CL1
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins