Learn More
ACROBiosystems Human Ephrin-B1 / EFNB1 Protein, His Tag
Marke: ACROBiosystems EF1-H5223-1MG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Ephrin-B1 (EFNB1) is also known as EFL-3, ELK ligand, EPH-related receptor tyrosine kinase ligand 2 (LERK-2), which belongs to the ephrin family. EFNB1 contains one ephrin RBD (ephrin receptor-binding) domain. EFNB1 binds to the receptor tyrosine kinases EPHB1 and EPHA1. EFNB1 binds to, and induce the collapse of, commissural axons/growth cones in vitro. EFNB1 may play a role in constraining the orientation of longitudinally projecting axons. EFNB1 interacts with GRIP1 and GRIP2.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 32-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_004420 | |
1 mg | |
Leu 28 - Gly 232 | |
EFNB1,EFL-3,ELK ligand,ELK-L,EPLG2,LERK2,CFND,CFNS | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
23.6 kDa | |
Human Ephrin-B1, His Tag (EF1-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 28 - Gly 232 (Accession # NP_004420). | |
-20°C | |
Ephrin-B1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |