Learn More
ACROBiosystems Biotinylated Human EGFRvIII Protein, Avitag™,His Tag
MALS verified
Marke: ACROBiosystems EGR-H82E0-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.The type III EGF deletion-mutant receptor (EGFRvIII) is the most common mutation and was first identified in primary human glioblastoma tumors; EGFR gene amplification is correlated with the structural rearrangement of the gene. The EGFRvIII gene has an in-frame deletion of 801 base pairs, corresponding to exons 2–7 in the mRNA, resulting in the deletion of amino acids 30-297 in the extracellular domain and the generation of a glycine at the fusion point.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 42.2 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001333870.1 | |
200 μg | |
Leu 25 - Ser 378 | |
EGFRvIII | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
42.2 kDa | |
Biotinylated Human EGFRvIII, Avitag,His Tag (EGR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1). | |
-20°C | |
EGFRvIII | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |