Learn More
ACROBiosystems Biotinylated Human LAIR2 / CD306 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems LA2-H82E4-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Leukocyte-associated immunoglobulin-like receptor 2 (LAIR2) is also known as CD antigen CD306. LAIR2 contains one Ig-like C2-type (immunoglobulin-like) domain. LAIR2 is a member of the immunoglobulin superfamily and was identified by its similarity to LAIR1, an inhibitory receptor present on mononuclear leukocytes. LAIR2 / CD306 is thought to be secreted and may help modulate mucosal tolerance.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.8 kDa. The protein migrates as 25-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002279.2 | |
25 μg | |
Gln 22 - Pro 152 | |
LAIR2,CD306 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |
17.8 kDa | |
Biotinylated Human LAIR-2, His,Avitag (LA2-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Pro 152 (Accession # Q6ISS4-1). | |
-20°C | |
LAIR-2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |