Learn More
ACROBiosystems Human Lipocalin-2 / NGAL Protein, His Tag
Marke:  ACROBiosystems LC2-H5222-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15867227
  • 335.90 € / 100 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Lipocalin-2 (LCN2) is also known as oncogene 24p3 or neutrophil gelatinase-associated lipocalin (NGAL), MSFI, The binding of lipocalin-2 to bacterial siderophores is important in the innate immune response to bacterial infection. Upon encountering invading bacteria the toll-like receptors on immune cells stimulate the synthesis and secretion of lipocalin-2. Secreted lipocalin-2 then limits bacterial growth by sequestering iron-containing siderophores. Lipocalin-2 also functions as a growth factor. LCN2 is strongly upregulated during inflammation and is upregulated by interleukin 1 (but not TNF alpha) in humans. There are indications that some forms of acne could be caused due to the gene not being transcribed, and that Isotretinoin corrects this. NFAT3 (NFATc4)NFAT by blocking the expression of LCN2 inhibits breast carcinoma cell motility. Recent studies have revealed that NGAL plays an important role in the physiopathology of chronic myeloid leukaemia (CML) mediated by BCR-ABL.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.4 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_005555.2
100 μg
Gln 21 - Gly 198
NGAL,LCN2,Lipocalin-2,24p3,MSFI
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
21.4 kDa
Human Lipocalin-2, His Tag (LC2-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Gly 198 (Accession # NP_005555.2).
-20°C
Lipocalin-2
Human
PBS, pH7.4
HEK293
Cytokines and Growth Factors