Learn More
ACROBiosystems Rabbit OX40 / TNFRSF4 / CD134 Protein, Fc Tag
Marke: ACROBiosystems OX0-R5254-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.3 kDa. As a result of different glycosylation, the protein migrates as 48 kDa and 68 kDa under reducing (R) condition, and 120-135 kDa under non-reducing (NRSpezifikation
47.3 kDa | |
Rabbit O x 40, Fc Tag (O x 0-R5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Gln 208 (Accession # O02764). | |
-20°C | |
OX40 | |
Rabbit | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
100 μg | |
Ala 19 - Gln 208 | |
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |