Learn More
ACROBiosystems Human SCARB1 / SR-B1 Protein, Fc Tag
Marke: ACROBiosystems SC1-H5258-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Scavenger receptor class B member 1 (SRB1) is also known as SR-BI, CD36 and LIMPII analogous 1 (CD36L1), CLA-1, is a member of the scavenger receptor family or CD36 family. CD36L1 is an integral membrane protein found in numerous cell types/tissues, including the liver and adrenal. SRB1 is receptor for different ligands such as phospholipids, cholesterol ester, lipoproteins, phosphatidylserine and apoptotic cells. CLA-1 facilitates the flux of free and esterified cholesterol between the cell surface and extracellular donors and acceptors, such as high-density lipoprotein (HDL) and to a lesser extent, apoB-containing lipoproteins and modified lipoproteins. SCARB1 is, along with CD81, the receptor for the entry of the Hepatitis C virus glycoprotein E2 in liver cells, and binding between SCARB1 and E2 was found to be independent of the genotype of the viral isolate. SRB1 plays an important role in the uptake of HDL cholesteryl ester.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.7 kDa. As a result of glycosylation, the protein migrates as 110-115 kDa under reducing (R) condition, and 220-230 kDa under non-reducing (NR) condition (SDSSpezifikation
NP_005496.4 | |
50 μg | |
Pro 33 - Tyr 443 | |
SCARB1,CD36L1,SRB1,CLA-1,SR-BI,HDLQTL6 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
72.7 kDa | |
Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 33 - Tyr 443 (Accession # NP_005496.4). | |
-20°C | |
SCARB1 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |