Learn More
ACROBiosystems Biotinylated Human CX3CL1 Protein, His Tag, ultra sensitivity (primary amine labeling)
Marke: ACROBiosystems CX1-H8221-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Fractalkine is also known as C-X3-C motif chemokine 1 (CX3CL1) and neurotactin. Fractalkine is a large cytokine protein of 373 amino acids, it contains multiple domains and is the only known member of the CX3C chemokine family. Fractalkine is found commonly throughout the brain, particularly in neural cells, and its receptor is known to be present on microglial cells. It has also been found to be essential for microglial cell migration. CX3CL1 is also up-regulated in the hippocampus during a brief temporal window following spatial learning, the purpose of which may be to regulate glutamate-mediated neurotransmission tone. This indicates a possible role for the chemokine in the protective plasticity process of synaptic scaling.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.5 kDa. The protein migrates as 50-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH01163 | |
25 μg | |
Gln 25 - Gln 341 | |
CX3CL1,Fractalkine | |
UltraLys | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
35.5 kDa | |
Biotinylated Human C x 3 CL1, His Tag, primary amine labeling (C x 1-H8221) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 341 (Accession # AAH01163). It is the biotinylated form of Human C x 3 CL1, His Tag (Cat. No.C x 1-H5221). | |
-20°C | |
CX3CL1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |