Learn More
ACROBiosystems Human DLL4 Protein, Fc Tag
Marke: ACROBiosystems DL4-H5259-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Delta-like protein 4 (DLL4) is also known as Drosophila Delta homolog 4 (Delta4). DLL4 contains one DSL domain and eight EGF-like domains. DLL4 is expressed in vascular endothelium. DLL4 involved in angiogenesis; negatively regulates endothelial cell proliferation and migration and angiogenic sprouting. DLL4 plays a role in the Notch signaling pathway. DLL4 activates Notch-1 and Notch-4.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 80.9 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_061947.1 | |
100 μg | |
Ser 27 - Pro 524 | |
DLL4,Delta4 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
80.9 kDa | |
Human DLL4, Fc Tag (DL4-H5259) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Pro 524 (Accession # NP_061947.1). | |
-20°C | |
DLL4 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |