Learn More
ACROBiosystems Biotinylated Human VEGF R3 / FLT4 Protein, His, Avitag™
Biotinylated Protein
Marke: ACROBiosystems FL4-H82E1-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Vascular endothelial growth factor receptor 3 (VEGF R3), also known as FLT-4, together with the other two members VEGFR1 (FLT-1) and VEGFR2 (KDR/Flk-1) are receptors for vascular endothelial growth factors (VEGF) and belong to the class III subfamily of receptor tyrosine kinases (RTKs). VEGF R3 mediates lymphangiogenesis in response to VEGF-C and VEGF-D. VEGF R3 is widely expressed in the early embryo but becomes restricted to lymphatic endothelia at later stages of development. It is likely that VEGF R3 may be important for lymph angiogenesis.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 88.2 kDa. The protein migrates as 115-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_002011.2 | |
200 μg | |
Tyr 25 - Ile 776 | |
Flt-4,FLT4,LMPH1A,PCLFLT41,VEGFR3,VEGFR-3,FLT-4,FLT-41,FLT41,PCL | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
88.2 kDa | |
Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Accession # P35916-1). | |
-20°C | |
VEGF R3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |