Learn More
ACROBiosystems Mouse GITR Ligand / TNFSF18 Protein, Fc Tag
Marke: ACROBiosystems GIL-M526X-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor ligand superfamily member 18 (TNFSF18) is also known as Glucocorticoid - induced TNF-related ligand (GITRL or GITR Ligand), Activation - inducible TNF - related ligand (AITRL), which belongs to the tumor necrosis factor family. TNFSF18 is expressed at high levels in the small intestine, ovary, testis, kidney and endothelial cells. TNFSF18 / GITRL is up-regulated after stimulation by bacterial lipopolysaccharides (LPS). TNFSF18 Can function as costimulator and lower the threshold for T-cell activation and T-cell proliferation. TNFSF18 / GITR Ligand is important for interactions between activated T-lymphocytes and endothelial cells. TNFSF18 also mediates activation of NF-kappa-B.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 41.4 kDa. As a result of glycosylation, the protein migrates as 45 kDa under reducing (R) condition, and 80-100 kDa under non-reducing (NR) condition (SDS-PAGE)Spezifikation
AAO89011 | |
100 μg | |
Thr 47 - Ser 173 | |
TNFSF18,AITRL,TL6,hGITRL,GITR Ligand | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
41.4 kDa | |
Mouse GITR Ligand, Fc Tag (GIL-M526x) is expressed from human 293 cells (HEK293). It contains AA Thr 47 - Ser 173 (Accession # AAO89011). | |
-20°C | |
GITR Ligand | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |