Learn More
ACROBiosystems Mouse ROR2 / NTRKR2 Protein, Fc Tag
Marke: ACROBiosystems RO2-M5253-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tyrosine-protein kinase transmembrane receptor ROR2 is also known as Neurotrophic tyrosine kinase, receptor-related 2 (NTRKR2), which belongs to the protein kinase superfamily and Tyr protein kinase family and ROR subfamily. ROR2 is a homodimer protein, which can binds YWHAB or interact with WTIP. ROR2 may be involved in the early formation of the chondrocytes. It seems to be required for cartilage and growth plate development.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.6 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
67.6 kDa | |
Mouse ROR2, Fc Tag (RO2-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 34 - Gly 403 (Accession # Q9Z138-1). | |
-20°C | |
ROR2 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
100 μg | |
Glu 34 - Gly 403 | |
ROR2,NTRKR2 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |