Learn More
ACROBiosystems Human UCH-L3 / UCHL3 Protein, His Tag
Marke: ACROBiosystems UC3-H5141-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Ubiquitin carboxyl-terminal hydrolase isozyme L3 (UCH-L3), a member of peptidase C12 family, is also known as ubiquitin thioesterase L3. Deubiquitinating enzyme (DUB) that controls levels of cellular ubiquitin through processing of ubiquitin precursors and ubiquitinated proteins. UCH-L3 is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of either ubiquitin or NEDD8. UCH-L3 indirectly increases the phosphorylation of IGFIR, AKT and FOXO1 and promotes insulin-signaling and insulin-induced adipogenesis. It is also required for stress-response retinal, skeletal muscle and germ cell maintenance. Furthermore, UCH-L3 may be involved in working memory and can hydrolyze UBB(+1), a mutated form of ubiquitin which is not effectively degraded by the proteasome and is associated with neurogenerative disorders.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27.0 kDa. The protein migrates as 27 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH18125.1 | |
100 μg | |
Glu 2 - Ala 230 | |
UCHL3,Ubiquitin thioesterase L3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
27.0 kDa | |
Human UCH-L3, His Tag (UC3-H5141) is expressed from E.coli cells. It contains AA Glu 2 - Ala 230 (Accession # P15374-1). | |
-20°C | |
UCH-L3 | |
Human | |
50 mM Tris, 150 mM NaCl, pH7.5 | |
E.coli | |
Enzymes |