Learn More
ACROBiosystems Human ALK-1 / ACVRL1 Protein, His Tag
Marke: ACROBiosystems AL1-H5227-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Serine/threonine-protein kinase receptor R3 is an enzyme that in humans is encoded by the ALK1 gene. ALK1 is a receptor in the TGF beta signaling pathway. ALK1 protein is a receptor in the TGF beta signaling pathway. It plays an important role in vascular development, remodeling, and pathologic angiogenesis, play a role in stabilizing angiogenic vessels and contribute to resistance to anti-VEGF therapies, ALK1 blockade may represent an effective therapeutic opportunity complementary to the current antiangiogenic modalities in the clinic. Recently, researcher found that, ALK1-Fc inhibited BMP9-mediated Id-1 expression in human umbilical vein endothelial cells and inhibited cord formation by these cells on a Matrigel substrate, in a chick chorioallantoic membrane assay, ALK1-Fc reduced vascular endothelial growth factor-, fibroblast growth factor-, and BMP10-mediated vessel formation, and ALK1-Fc treatment reduced tumor burden in mice receiving orthotopic grafts of MCF7 mammary adenocarcinoma cells.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.5 kDa. The protein migrates as 22-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000011.2 | |
100 μg | |
Asp 22 - Gln 118 | |
ACVRL1,ACVRLK1,ALK-1,HHT,HHT2,ORW2,SKR3,TSR-I | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>97% as determined by SDS-PAGE |
11.5 kDa | |
Human ALK-1, His Tag (AL1-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Gln 118 (Accession # NP_000011.2). | |
-20°C | |
ALK-1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |