Learn More
ACROBiosystems Biotinylated Human ANGPTL7 / CDT6 Protein, Mouse IgG2a Fc, Avitag™
Biotinylated Protein
Marke: ACROBiosystems AN7-H82F3-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Angiopoietin-like 7 (ANGPTL7) is also known as Corneal-Derived Transcript 6 (CDT6), and is a secreted glycoprotein that is structurally related to the angiopoietins. It is expressed in the corneal stroma, trabecular meshwork, and sclera and is elevated in glaucoma aqueous humor. Recently, ANGPTL proteins, including ANGPTL-2, -3, -4, -5 and -7, are identified as growth factors in combination with SCF, Thrombopoietin, IGF-II and FGF acidic that enhance the expansion and engraftment of human and mouse hematopoietic stem cells.
This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 65.8 kDa. The protein migrates as 66 kDa and 75-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_066969.1 | |
25 μg | |
Gln 27 - Pro 346 | |
ANGPTL7,CDT6 | |
Precision Avi | |
Mouse IgG Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
65.8 kDa | |
Biotinylated Human ANGPTL7, Mouse IgG2a Fc,Avitag (AN7-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Pro 346 (Accession # O43827-1). | |
-20°C | |
ANGPTL7 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |