Learn More
ACROBiosystems Human CD14 Protein, Fc Tag
Marke: ACROBiosystems CD4-H5252-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Cluster of differentiation 14 (CD14), is a cell surface glycoprotein, and is a is a component of the innate immune system. CD14 is a myelomonocytic differentiation antigen preferentially expressed on monocytes, macrophages, and activated granulocytes. CD14 exists in two forms. Either it is anchored into the membrane by a glycosylphosphatidylinositol tail (mCD14) or it appears in a soluble form (sCD14). Soluble CD14 either appears after shedding of mCD14 (48 kDa) or is directly secreted from intracellular vesicles (56 kDa). CD14 acts as a co-receptor (along with the Toll-like receptor TLR 4 and MD-2) for the detection of bacterial lipopolysaccharide (LPS). CD14 can bind LPS only in the presence of lipopolysaccharide-binding protein (LBP). CD14 has been proposed to be involved in various biological processes, including transportation of other lipids, cell-cell interaction during different immune responses, as well as recognition of apoptotic cells. Although LPS is considered its main ligand, CD14 also recognizes other pathogen-associated molecular patterns. CD14+ cells are monocytes that can differentiate into a host of different cells. CD14 has been shown to interact with Lipopolysaccharide-binding protein.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.2 kDa. The protein migrates as 75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000582.1 | |
100 μg | |
Thr 20 - Cys 352 | |
CD14 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
62.2 kDa | |
Human CD14, Fc Tag (CD4-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Cys 352 (Accession # P08571-1). | |
-20°C | |
CD14 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |