Learn More
ACROBiosystems Human E-Selectin / CD62E Protein, His Tag
Marke: ACROBiosystems CDE-H5222-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
E-Selectin is also known as CD62 antigen-like family member E (CD62E), endothelial-leukocyte adhesion molecule 1 (ELAM-1), or leukocyte-endothelial cell adhesion molecule 2 (LECAM2), a member of the Selectin family, is a 107-115 kDa cell surface glycoprotein. It is transiently expressed on vascular endothelial cells in response to IL-1β and TNFα. E selectin has a cassette structure: an N-terminal, C-type lectin domain, an EGF (epidermal-growth-factor)-like domain, 6 Sushi domain (SCR repeat) units, a transmembrane domain (TM) and an intracellular cytoplasmic tail (cyto). During inflammation, E-selectin plays an important part in recruiting leukocytes to the site of injury. The local release of cytokines IL-1 and TNF-α by damaged cells induces the over-expression of E-selectin on endothelial cells of nearby blood vessels. E-selectin mediates the adhesion of tumor cells to endothelial cells, by binding to E-selectin ligands expressed by neutrophils, monocytes, eosinophils, memory-effector T-like lymphocytes, natural killer cells or cancer cells. Furthermore, a number of studies have reported that levels of E-Selectin may be elevated in subjects with a variety of pathological conditions.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.5 kDa. The protein migrates as 110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_000441.2 | |
200 μg | |
Trp 22 - Pro 556 | |
SELE,RP1-117P20.2,CD62E,ELAM1,ESEL,LECAM2,E-Selectin | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
59.5 kDa | |
Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession # NP_000441.2). | |
-20°C | |
E-Selectin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |