missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse CD40 Ligand / TNFSF5 Protein, Fc Tag

Marke:  ACROBiosystems CDL-M526X-100UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15886427

  • 359.89 € / 100 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CD40 ligand is also known as CD40L, CD154, TNFSF5 and T-cell antigen Gp39, is a single-pass type I I membrane protein which belongs to the TNF superfamily of molecules. CD40 ligand is expressed predominantly on activated CD4+ T lymphocytes, and also found in other types of cells, including platelets, mast cells, macrophages, basophils, NK cells, B lymphocytes, as well as non-haematopoietic cells (smooth muscle cells, endothelial cells, and epithelial cells). Although all monomeric, dimeric and trimeric forms of soluble CD40 ligand can bind to CD40, the trimeric form of soluble CD40 ligand has the most potent biological activity through oligomerization of cell surface CD40, a common feature of TNF receptor family members.CD40 ligand binds to CD40 on antigen-presenting cells (APC), which leads to many effects depending on the target cell type. In general, CD40 ligand plays the role of a costimulatory molecule and induces activation in APC in association with T cell receptor stimulation by MHC molecules on the APC. In total CD40 ligand has three binding partners: CD40, α5β1 integrin and αIIbβ3. CD40 ligand regulates B cell function by engaging CD40 on the B cell surface. A defect in this gene results in an inability to undergo immunoglobulin class switch and is associated with hyper IgM syndrome.

This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 42.9 kDa. As a result of glycosylation, the protein migrates as 52 kDa under reducing (R) condition, and 90-116 kDa under non-reducing (NR) condition (SDS-PAGE)
Spezifikation

Spezifikation

42.9 kDa
Mouse CD40 Ligand, Fc Tag (CDL-M526x) is expressed from human 293 cells (HEK293). It contains AA Gly 115 - Leu 260 (Accession # P27548-1).
-20°C
CD40 Ligand
Mouse
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Immune Checkpoint Proteins
100 μg
Gly 115 - Leu 260
CD40LG,CD154,CD40L,HIGM1,IGM,IMD3,T-BAM,TNFSF5,TRAP,gp39
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote