missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human IL-1RL1 / ST2 Protein, Fc Tag

Marke:  ACROBiosystems IL1-H5250-1MG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15887017

  • 988.62 € / 1 Milligramm

Voraussichtliches Versanddatum: 09-08-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Interleukin 1 receptor-like 1 (IL1RL1) is also known as ST2, DER4, FIT-1, IL33R, ST2L, ST2V, which belongs to the interleukin-1 receptor family. IL1RL1 contains three Ig-like C2-type (immunoglobulin-like) domains and one TIR domain. IL1RL1 is receptor for interleukin-33 (IL-33), its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. IL1RL1 possibly involved in helper T-cell function. IL1RL1 can interact with MYD88, IRAK1, IRAK4, and TRAF6.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 61.0 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

NP_003847
HEK293
Human
61.0 kDa
1 mg
Lys 19 - Pro 323
IL1RL1,DER4,FIT-1,IL33R,ST2,ST2L,ST2V,T1
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human IL-1 RL1, Fc Tag (IL1-H5250) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Pro 323 (Accession # NP_003847).
-20°C
IL-1 RL1
Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate