Learn More
ACROBiosystems Human IL-20 R beta Protein, Fc Tag
Marke: ACROBiosystems ILB-H5258-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Interleukin-20 receptor subunit beta (IL20RB) is also known as IL-20R2, DIRS1, which belongs to the type II cytokine receptor family. IL20RB / IL-20R2 contains two fibronectin type-III domains. IL20RB / IL-20R2 is widely expressed with highest levels in skin and testis. IL20RB can be fused with IL20RA or IL22RA1 forming heterodimer. The IL20RA/IL20RB dimer is a receptor for IL19, IL20 and IL24. The IL22RA1/IL20RB dimer is a receptor for IL20 and IL24.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.1 kDa. As a result of glycosylation, the protein migrates as 50 kDa under reducing (R) condition, and 100 kDa under non-reducing (NR) condition (SDS-PAGE).Spezifikation
AAH63696 | |
100 µg | |
Asp 30 - Ala 230 | |
IL20RB,IL-20R2,IL-20 R-beta,DIRS1,FNDC6 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
49.1 kDa | |
Human IL-20 R beta, Fc Tag (ILB-H5258) is expressed from human 293 cells (HEK293). It contains AA Asp 30 - Ala 230 (Accession # AAH63696). | |
-20°C | |
IL-20 R beta | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |