Learn More
ACROBiosystems Human IL-1 RAcP / IL-1 R3 Protein, Fc Tag
Marke: ACROBiosystems ILP-H5256-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Interleukin-1 receptor accessory protein (IL1RAP or IL-1RAcP) is also known as Interleukin-1 receptor 3 (IL-1R-3 or IL-1R3), C3orf13, which belongs to the interleukin-1 receptor family. IL1RAP contains three Ig-like C2-type (immunoglobulin-like) domains and one TIR domain. IL1RAP is detected in liver, skin, placenta, thymus and lung. IL-1R3 / IL1RAP is coreceptor with IL1R1, associates with IL1R1 bound to IL1B to form the high affinity interleukin-1 receptor complex which mediates interleukin-1-dependent activation of NF-kappa-B and other pathways.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 65.7 kDa. The protein migrates as 75-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
65.7 kDa | |
Human IL-1 RAcP, Fc Tag (ILP-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Glu 359 (Accession # Q9NPH3). | |
-20°C | |
IL-1 RAcP | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
100 μg | |
Ser 21 - Glu 359 | |
IL1RAP,IL-1 RAcP,C3orf13,IL1R3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |