Learn More
ACROBiosystems Human CD4 / LEU3 Protein, His Tag
Marke:  ACROBiosystems LE3-H5228-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15887237
  • 419.88 € / 200 Mikrogramm
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
T-cell surface glycoprotein CD4 is also known as T-cell surface antigen T4/Leu-3. CD4 contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. CD4 is accessory protein for MHC class-II antigen/T-cell receptor interaction. CD4 induces the aggregation of lipid rafts. CD4 is a primary receptor used by HIV-1 to gain entry into host T cells. HIV infection leads to a progressive reduction of the number of T cells possessing CD4 receptors. Therefore, medical professionals refer to the CD4 count to decide when to begin treatment for HIV-infected patients.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.5 kDa. The protein migrates as 47-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH25782
200 μg
Lys 26 - Trp 390
CD4,CD4mut,LEU3
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
41.5 kDa
Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession # AAH25782).
-20°C
CD4
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens