Learn More
ACROBiosystems Human Reg4 Protein, His Tag
Marke: ACROBiosystems RE4-H5221-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Regenerating islet-derived protein 4 (REG4) is also known as Gastrointestinal secretory protein (GISP), REG-like protein (RELP). REG4 contains one C-type lectin domain. REG4 has calcium-independent lectin displaying mannose-binding specificity and able to maintain carbohydrate recognition activity in an acidic environment. REG4 may be involved in inflammatory and metaplastic responses of the gastrointestinal epithelium.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH17089 | |
100 μg | |
Asp 23 - Pro 158 | |
REG4,GISP,REG-IV,RELP | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
16.7 kDa | |
Human Reg4, His Tag (RE4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Pro 158 (Accession # AAH17089). | |
-20°C | |
Reg4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |