Learn More
ACROBiosystems Human TFPI-2 Protein, His Tag
Marke: ACROBiosystems TF2-H5227-50UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Tissue Factor Pathway Inhibitor 2 (TFPI2) is also known as placental protein 5 (PP5) and retinal pigment epithelial cell factor 1 (REF1), is a member of the Kunitz-type serine proteinase inhibitor family, is a structural homologue of tissue factor pathway inhibitor (TFPI). TFPI2 is highly abundant in the full-term placenta and widely expressed in umbilical vein endothelial cells, liver, placenta, heart, pancreas, and maternal serum at advanced pregnancy. TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. Inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa and has no effect on thrombin. Reduced synthesis of TFPI-2 has been related to numerous pathophysiological processes such as inflammation, angiogenesis, atherosclerosis, retinal degeneration and tumor growth / metastasis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.7 kDa. The protein migrates as 26-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_006519.1 | |
50 μg | |
Asp 23 - Lys 213 | |
TFPI2,PP5,REF1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
22.7 kDa | |
Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Lys 213 (Accession # NP_006519.1). | |
-20°C | |
TFPI-2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |