Learn More
ACROBiosystems Biotinylated Human Transferrin R / CD71 Protein, His Tag, ultra sensitivity (primary amine labeling)
Marke: ACROBiosystems TFR-H8243-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia & lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 76.0 kDa. The protein migrates as 75-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH01188.1 | |
25 μg | |
Cys 89 - Phe 760 | |
CD71,TFRC,TFR,sTfR,TfR1,Trfr,T9,p90 | |
UltraLys | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
76.0 kDa | |
Biotinylated Human Transferrin R, His Tag, primary amine labeling is expressed from human HEK293 cells. It contains AA Cys 89 - Phe 760 (Accession # AAH01188.1). It is the biotinylated form of Human Transferrin R protein, His Tag (Cat. No. CD1-H5243). | |
-20°C | |
Transferrin R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |