Learn More
ACROBiosystems Human Frizzled-7 / FZD7 Protein, Fc Tag
Marke: ACROBiosystems FZ7-H5258-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Frizzled-7 (FZD7) is also known as FzE3, which belongs to the G-protein coupled receptor Fz/Smo family. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. FZD7 contains one FZ (frizzled) domain. FZD7 is receptor for Wnt proteins. FZD7 may be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. FZD7 interacts with MAGI3 and DVL1.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.3 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH15915 | |
100 μg | |
Gln 33 - Leu 185 | |
FZD7,Frizzled-7,FzE3,Fz-7,hFz7 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
43.3 kDa | |
Human Frizzled-7, Fc Tag (FZ7-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 33 - Leu 185 (Accession # AAH15915). | |
-20°C | |
Frizzled-7 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |