Learn More
ACROBiosystems Human IFN-gamma R1 / IFNGR1 Protein, Fc Tag
Marke:  ACROBiosystems IF1-H5254-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15896967
  • 347.90 € / 200 Mikrogramm
Verfügbar ab: 09-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Interferon gamma receptor 1 (IFNGR1) is also known as CD antigen CD119, which belongs to the type II cytokine receptor family. IFNGR1 contains two fibronectin type-III domains and two Ig-like C2-type (immunoglobulin-like) domains. IFNGR1 / CD119 is receptor for interferon gamma. Two receptors bind one interferon gamma (IFNG) dimer.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.4 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
AAH05333
200 μg
Glu 18 - Gly 245
IFNGR1,CD119,CDw119,IFNGR,IFN-gamma-R1
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
52.4 kDa
Human IFN-gamma R1, Fc Tag (IF1-H5254) is expressed from human 293 cells (HEK293). It contains AA Glu 18 - Gly 245 (Accession # AAH05333).
-20°C
IFN-gamma R1
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens