Learn More
ACROBiosystems Biotinylated Human VEGF R2 / KDR Protein, Avitag™,His Tag
Biotinylated Protein
Marke: ACROBiosystems KDR-H82E5-25UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Kinase insert domain receptor (KDR) is also known as CD309, FLK1, VEGFR, VEGFR2, and is one of the subtypes of VEGFR. VEGF receptors are receptors for vascular endothelial growth factor (VEGF). There are three main subtypes of VEGFR, numbered 1, 2 and 3. The VEGF receptors have an extracellular portion consisting of 7 immunoglobulin-like domains, a single transmembrane spanning region and an intracellular portion containing a split tyrosine-kinase domain. VEGF-A binds to VEGFR-1 (Flt-1) and VEGFR-2 (KDR/Flk-1). VEGFR-2 appears to mediate almost all of the known cellular responses to VEGF.The function of VEGFR-1 is less well defined, although it is thought to modulate VEGFR-2 signaling. Another function of VEGFR-1 may be to act as a dummy/decoy receptor, sequestering VEGF from VEGFR-2 binding (this appears to be particularly important during vasculogenesis in the embryo). In addition, VEGFR2 is able to interact with HIV-1 extracellular Tat protein upon VEGF activation, and seems to enhance angiogenesis in Kaposi's sarcoma lesions.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 85.9 kDa. The protein migrates as 120-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAI31823 | |
25 μg | |
Ala 20 - Glu 764 | |
KDR,CD309,FLK1,VEGFR,VEGFR2 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
85.9 kDa | |
Biotinylated Human VEGF R2, Avitag,His Tag (KDR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823). | |
-20°C | |
VEGF R2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |