Learn More
ACROBiosystems Marmoset LAG-3 / CD223 Protein, His Tag
Marke: ACROBiosystems LA3-M52HA-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Lymphocyte activation gene 3 protein (LAG3) is also known as CD antigen CD223 and protein FDC, which belongs to immunoglobulin (Ig) superfamily and contains 4 extracellular Ig-like domains. The LAG3 gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG3 to CD4. LAG3 /CD223 involved in lymphocyte activation. LAG3 /CD223 binds to HLA class-II antigens.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.0 kDa. The protein migrates as 57 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
XP_002752325.1 | |
100 μg | |
Ala 18 - His 449 | |
LAG3,CD223,FDC | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
48.0 kDa | |
Marmoset LAG-3, His Tag (LA3-M52Ha) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession # XP_002752325.1). | |
-20°C | |
LAG-3 | |
Marmoset | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |