Learn More
ACROBiosystems Zika virus NS1 Protein, His Tag
Marca: ACROBiosystems NS1-Z5222-50UG
Detalles adicionales : Netto-Gewicht : 0.00001kg
Descripción
Zika virus (ZIKV) is a member of the virus family Flaviviridae and the genus Flavivirus, transmitted by daytime-active Aedes mosquitoes, such as A. aegypti and A. albopictus. Its name comes from the Zika Forest of Uganda, where the virus was first isolated in 1947. Zika virus is related to dengue, yellow fever, Japanese encephalitis, and West Nile viruses. The infection, known as Zika fever, often causes no or only mild symptoms, similar to a mild form of dengue fever. It is treated by rest. Since the 1950s, it has been known to occur within a narrow equatorial belt from Africa to Asia. As of 2016, the illness cannot be prevented by drugs or vaccines. As of February 2016, there is evidence that Zika fever in pregnant women is associated with abnormal brain development in their fetuses through mother-to-child transmission of the virus, which may result in miscarriage or microcephaly.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 42.0 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE).Especificaciones
42.0 kDa | |
Zika virus NS1, His Tag (NS1-Z5222) is expressed from human 293 cells (HEK293). It contains AA Val 796 - Ser 1148 (Accession # ALU33341). | |
-20°C | |
NS1 | |
Zika virus | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
50 μg | |
Val 796 - Ser 1148 | |
NS1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |