Learn More
ACROBiosystems Human Nectin-3 / CD113 Protein, Fc Tag
Marke: ACROBiosystems PV3-H5255-100UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
Poliovirus receptor-related 3 (PVRL3), also known as nectin-3 and CD113, is a human protein of the immunoglobulin superfamily which forms part of adherens junctions. Nectins are immunoglobulin-like adhesion molecules that interact with afadin (AF6; MIM 159559). Afadin is an actin filament-binding protein that connects nectins to the actin cytoskeleton. The nectin-afadin system organizes adherens junctions cooperatively with the cadherin system in epithelial cells. PVRL3 plays a role in cell-cell adhesion through heterophilic trans-interactions with nectin-like proteins or nectins, such as trans-interaction with PVRL2/nectin-2 at Sertoli-spermatid junctions. Furthermore, PVRL3 induces endocytosis-mediated down-regulation of PVR from the cell surface, resulting in reduction of cell movement and proliferation.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.3 kDa. The protein migrates as 80-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
Q9NQS3 | |
100 μg | |
Gly 58 - Asp 400 | |
CD113,Nectin-3,PVRL3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
64.3 kDa | |
Human Nectin-3, Fc Tag (PV3-H5255) is expressed from human 293 cells (HEK293). It contains AA Gly 58 - Asp 400 (Accession # Q9NQS3). | |
-20°C | |
Nectin-3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |