Learn More
Alfa Aesar™ 16-Hydroxyhexadecansäure, 98 %
Marke: Alfa Aesar™ A17845.06
Weitere Details : CAS : 506-13-8 Netto-Gewicht : 0.00500kg
Menge | 5g |
---|
Chemische Identifikatoren
506-13-8 | |
272.429 | |
UGAGPNKCDRTDHP-UHFFFAOYSA-N | |
10466 | |
16-Hydroxyhexadecansäure |
C16H32O3 | |
MFCD00002750 | |
juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid | |
CHEBI:55328 | |
C(CCCCCCCC(=O)O)CCCCCCCO |
Spezifikation
16-Hydroxyhexadecanoic acid | |
5g | |
C16H32O3 | |
MFCD00002750 | |
juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid | |
C(CCCCCCCC(=O)O)CCCCCCCO | |
272.429 | |
CHEBI:55328 | |
98% |
98°C to 100°C | |
506-13-8 | |
HO(CH2)15CO2H | |
1783998 | |
UGAGPNKCDRTDHP-UHFFFAOYSA-N | |
16-Hydroxyhexadecansäure | |
10466 | |
272.43 |