missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems PE-Labeled Human SIRP alpha / CD172a Protein, Fc Tag (recommended for neutralizing assay)

Marke:  ACROBiosystems SIA-HP252-50TESTS

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16358731

  • 383.89 € / 50 Tests
Verfügbar ab: 08-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Tyrosine-protein phosphatase non-receptor type substrate 1 (SHPS1) is also known as CD172 antigen-like family member A (CD172a), Macrophage fusion receptor, MyD-1 antigen, Signal-regulatory protein alpha (SIRPA or SIRP alpha) or p84, is a member of the SIRP family, and also belongs to the immunoglobulin superfamily. SIRP alpha is Ubiquitous and highly expressed in brain. SIRPA / CD172a is immunoglobulin-like cell surface receptor for CD47 and acts as docking protein and induces translocation of PTPN6, PTPN11 and other binding partners from the cytosol to the plasma membrane. SIRPA / SHPS-1 supports adhesion of cerebellar neurons, neurite outgrowth and glial cell attachment and may play a key role in intracellular signaling during synaptogenesis and in synaptic function By similarity. SIRPA / MyD1 involved in the negative regulation of receptor tyrosine kinase-coupled cellular responses induced by cell adhesion, growth factors or insulin and mediates negative regulation of phagocytosis, mast cell activation and dendritic cell activation. CD47 binding prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 65.7 kDa.
Spezifikation

Spezifikation

NP_001035111.1
50 tests
Glu 31 - Arg 370
SHPS1,SIRPA,CD172A,BIT,MFR,MYD1,P84,PTPNS1
PE Labeled
Fc Tag
HEK293
Fluorescent labeled protein
65.7 kDa
PE-Labeled Human SIRP alpha, Fc Tag (recommended for neutralizing assay) (Cat. No. SIA-HP252) is produced via conjugation of PE to Human SIRP alpha, Fc tag under optimal conditions. Human SIRP alpha, Fc tag is expressed in HEK293 cells containing AA Glu 31 - Arg 370 (Accession # P78324-1).
-20°C, avoid light
SIRP alpha
Human
PBS, 0.5% BSA, pH7.4
Powder
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote