missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human CD2 / SRBC Protein, Fc Tag

Marke:  ACROBiosystems CD2-H5258-1mg

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16407269

  • 2421.11 € / 1 Milligramm

Voraussichtliches Versanddatum: 19-10-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

T-cell surface antigen CD2 is also known as Erythrocyte receptor, LFA-2, LFA-3 receptor, Rosette receptor, T-cell surface antigen T11/Leu-5 and SRBC, is a single-pass type I membrane protein found on the surface of T cells and natural killer (NK) cells. CD2 is a member of the immunoglobulin superfamily. CD2 / SRBC contains 1 Ig-like C2-type (immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain. CD2 / SRBC interacts with other adhesion molecules, such as lymphocyte function-associated antigen-3 (LFA-3 / CD58) in humans, or CD48 in rodents, which are expressed on the surfaces of other cells. In addition to its adhesive properties, CD2 also acts as a co-stimulatory molecule on T and NK cells. CD2 is a specific marker for T cells and NK cells, and can therefore be used in immunohistochemistry to identify the presence of such cells in tissue sections.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.7 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

AAH33583
HEK293
Human
47.7 kDa
1 mg
Lys 25 - Asp 209
CD2,SRBC,LFA-2,T11
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human CD2, Fc Tag (CD2-H5258) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accession # AAH33583).
-20°C
CD2
Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate