missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human CD229 / SLAMF3 Protein, His Tag

Marke:  ACROBiosystems CD9-H52H6-100ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16407349

  • 302.64 € / 100 Mikrogramm

Voraussichtliches Versanddatum: 07-10-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CD229, also known as Ly9 and SLAMF3, is a 120 kDa type I transmembrane glycoprotein in the SLAM subgroup of the CD2 family. Signaling lymphocyte activation molecule (SLAM) family receptors are critically involved in modulating innate and adaptive immune responses. CD229 is expressed on T and B cells, thymocytes, and more weakly on NK cells. Homophilic binding between CD229 molecules is mediated by the N-terminal Ig-like domain.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.7 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

NP_002339.2
HEK293
Human
46.7 kDa
100 µg
Lys 48 - Lys 454
SLAMF3,LY9,Ly-9,hly9,Ly9
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>90% as determined by SDS-PAGE
Human CD229, His Tag (CD9-H52H6) is expressed from human 293 cells (HEK293). It contains AA Lys 48 - Lys 454 (Accession # Q9HBG7-1).
-20°C
CD229
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate